3,3'-Dimethyltriphenylamine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N,N-Di-m-tolylaniline, Bis(3-methylphenyl)phenylamine, Phenyl(di-m-tolyl)amine
D4755-1G
D4755-5G
TCIAD4755-1GEA
460
DKK
TCIAD4755-1G
TCIAD4755-5G
3,3'-Dimethyltriphenylamine ≥98.0% (by GC)
3,3'-Dimethyltriphenylamine
Formel:
C₂₀H₁₉N |
MDL Number:
MFCD23115649 CAS-nummer: 13511-11-0 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...