1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol, 1-[(3,3-Diphenylpropyl)methylamino]-2-methyl-2-propanol

D4713-5G D4713-25G
TCIAD4713-5GEA 378 DKK
TCIAD4713-5G TCIAD4713-25G
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol ≥98.0% (by GC, titration analysis)
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol
Formel: C₂₀H₂₇NO
Molekylvægt: 297.43 g/mol
Kogepunkt: 145°C
MDL Number: MFCD07782118
CAS-nummer: 100442-33-9

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR