2-[[2-(Dimethylamino)ethyl]methylamino]ethanol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4680-25ML D4680-100ML
TCIAD4680-25MLEA 296 DKK
TCIAD4680-25ML TCIAD4680-100ML
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol ≥97.0% (by GC, titration analysis)
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol
Formel: C₇H₁₈N₂O
Kogepunkt: 88°C
Flammepunkt: 87°C
MDL Number: MFCD00010387
CAS-nummer: 2212-32-0
EINECS: 218-658-4

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR