2,3-Diamino-5-bromo-6-methylpyridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4658-1G D4658-5G
TCIAD4658-1GEA 501 DKK
TCIAD4658-1G TCIAD4658-5G
2,3-Diamino-5-bromo-6-methylpyridine ≥98.0% (by GC)
2,3-Diamino-5-bromo-6-methylpyridine
Formel: C₆H₈BrN₃
MDL Number: MFCD00160167
CAS-nummer: 59352-90-8
EINECS: 261-714-8

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR