1,4-Dioxan-2-one ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: p-Dioxanone

D4644-5G D4644-25G
TCIAD4644-5GEA 657 DKK
TCIAD4644-5G TCIAD4644-25G
1,4-Dioxan-2-one ≥98.0% (by GC)
1,4-Dioxan-2-one
Formel: C₄H₆O₃
Molekylvægt: 102.09 g/mol
Kogepunkt: 215°C
Smeltepunkt: 28°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00215855
CAS-nummer: 3041-16-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR