9,9-Di(p-tolyl)fluorene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D4610-1G D4610-5G
TCIAD4610-1GEA 731 DKK
TCIAD4610-1G TCIAD4610-5G
9,9-Di(p-tolyl)fluorene ≥98.0% (by GC)
9,9-Di(p-tolyl)fluorene
Formel: C₂₇H₂₂
MDL Number: MFCD23115707
CAS-nummer: 54941-50-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR