4-Dibenzofuranboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D4567-1G
D4567-5G
TCIAD4567-1GEA
181
DKK
TCIAD4567-1G
TCIAD4567-5G
4-Dibenzofuranboronic acid (contains varying amounts of Anhydride)
4-Dibenzofuranboronic acid
Formel:
C₁₂H₉BO₃ Smeltepunkt: 258°C |
MDL Number:
MFCD00092336 CAS-nummer: 100124-06-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | gt. 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...