2,6-Dichlorbenzylamin ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D4521-5G D4521-25G
TCIAD4521-5GEA 1700 DKK
TCIAD4521-5G TCIAD4521-25G
2,6-Dichlorbenzylamin ≥97.0% (by GC, titration analysis)
2,6-Dichlorbenzylamin
Formel: C₇H₇Cl₂N
Kogepunkt: 244°C
MDL Number: MFCD00047928
CAS-nummer: 6575-27-5
UN nummer: 2735
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR