2,4-Dichlor-α-methylbenzylalkohol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2,4-Dichlorophenyl methyl carbinol, 2,4-Dichloro-alpha-methylbenzyl alcohol, 1-(2,4-Dichlorophenyl)ethyl alcohol, 1-(2,4-Dichlorophenyl)ethanol
D4508-5G
D4508-25G
TCIAD4508-5GEA
427
DKK
TCIAD4508-5G
TCIAD4508-25G
2,4-Dichlor-α-methylbenzylalkohol ≥98.0% (by GC)
2,4-Dichlor-α-methylbenzylalkohol
Formel:
C₈H₈Cl₂O Kogepunkt: 126°C Flammepunkt: 108°C |
MDL Number:
MFCD00004511 CAS-nummer: 1475-13-4 EINECS: 210-019-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...