4,5-Dimethoxy-1-benzocyclobutenecarbonitrile ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile, 4,5-Dimethoxy-1-cyanobenzocyclobutane, 1-Cyano-4,5-dimethoxybenzocyclobutene

D4485-1G D4485-5G
TCIAD4485-1GEA 682 DKK
TCIAD4485-1G TCIAD4485-5G
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile ≥98.0% (by HPLC, total nitrogen)
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile
Formel: C₁₁H₁₁NO₂
MDL Number: MFCD01846132
CAS-nummer: 35202-54-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR