Dess-Martin periodinane 8 - 12% in dichloromethane

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

D4477-25ML D4477-250ML
TCIAD4477-25MLEA 665 DKK
TCIAD4477-25ML TCIAD4477-250ML
Dess-Martin periodinane 8 - 12% in dichloromethane
Dess-Martin periodinane
Formel: C₁₃H₁₃IO₈
Molekylvægt: 424.15 g/mol
Flammepunkt: –11°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00130127
CAS-nummer: 87413-09-0
UN nummer: 3139
ADR: 5.1 III
Merck Index: 14,02933
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa18620f1b3b69de396787de83bada1577ceaf4e&vid=8993fe4100bf99a12786b3d71eb33028b9061f57&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Concentration (Iodometric titration) 8.0 to 12.0 w/w%

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR