N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylanilinium tetrakis (pentafluorophenyl)borate, N,N-Dimethylanilinium tetra(pentafluorophenyl)borate, N,N-Dimethylbenzenaminium tetrakis(perfluorophenyl)borate

D4404-200MG D4404-1G
TCIAD4404-200MGEA 444 DKK
TCIAD4404-200MG TCIAD4404-1G
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate
Formel: C₃₂H₁₂BF₂₀N
MDL Number: MFCD12913898
CAS-nummer: 118612-00-3
EINECS: 422-050-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR