2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D4385-1ML
D4385-5ML
TCIAD4385-1MLEA
378
DKK
TCIAD4385-1ML
TCIAD4385-5ML
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) ≥95.0% (by GC)
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers)
Formel:
C₁₅H₂₈O Kogepunkt: 134°C |
MDL Number:
MFCD27976821 CAS-nummer: 23790-39-8 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % (total of isomers |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...