Dibenzo[a,e]cyclooctene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: DBCOT, Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene

D4338-200MG D4338-1G
TCIAD4338-200MGEA 747 DKK
TCIAD4338-200MG TCIAD4338-1G
Dibenzo[a,e]cyclooctene ≥97.0% (by GC)
Dibenzo[a,e]cyclooctene
Formel: C₁₆H₁₂
Kogepunkt: 130°C
Opbevaringstemperatur: Køleskab
CAS-nummer: 262-89-5

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 105.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR