(3-(Dimethylamino)propyl)trimethoxysilan ≥96.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(N,N-Dimethylaminopropyl)trimethoxysilane, N,N-Dimethyl-3-(trimethoxysilyl)propylamine, N,N-Dimethyl-3-(trimethoxysilyl)propan-1-amine, Trimethoxy(3-(dimethylamino)propyl)silane, (3-Dimethylaminopropyl)trimethoxysilane, [3-(N,N-Dimethylamino)propyl]trimethoxysilane
D4328-5ML
D4328-25ML
TCIAD4328-5MLEA
263
DKK
TCIAD4328-5ML
TCIAD4328-25ML
(3-(Dimethylamino)propyl)trimethoxysilan ≥96.0% (by GC, titration analysis)
(3-(Dimethylamino)propyl)trimethoxysilane
Formel:
(H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃ Molekylvægt: 207.35 g/mol Kogepunkt: 42°C Flammepunkt: 99°C |
MDL Number:
MFCD00053674 CAS-nummer: 2530-86-1 EINECS: 219-786-3 UN nummer: 1760 ADR: 8 III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...