Dipropylenglycol dimethylether (mix af isomere) ≥94.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dipropylene glycol dimethyl ether
D4283-25ML
D4283-500ML
TCIAD4283-25MLEA
140
DKK
TCIAD4283-25ML
TCIAD4283-500ML
Dipropylenglycol dimethylether (mix af isomere) ≥94.0% (by GC)
Dipropylenglycol dimethylether (mix af isomere)
Formel:
C₈H₁₈O₃ Flammepunkt: 65°C |
MDL Number:
MFCD00210047 CAS-nummer: 111109-77-4 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...