(S)-3-[1-(Dimethylamino)ethyl]phenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-3-(1-(Dimethylamino)ethyl)phenol, NAP-226-90

D4258-5G D4258-1G
TCIAD4258-5GEA 476 DKK
TCIAD4258-5G TCIAD4258-1G
(S)-3-[1-(Dimethylamino)ethyl]phenol ≥98.0% (by GC, titration analysis)
(S)-3-[1-(Dimethylamino)ethyl]phenol
Formel: C₁₀H₁₅NO
Molekylvægt: 165.24 g/mol
MDL Number: MFCD06656491
CAS-nummer: 139306-10-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C
Specific rotation [a]20/D -53.0 to -58.0 deg (C=1、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR