Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic Acid Diethyl Ester, 9-[2-(Diethoxyphosphorylmethoxy)ethyl]adenine

D4256-1G D4256-5G
TCIAD4256-1GEA 460 DKK
TCIAD4256-1G TCIAD4256-5G
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate
Formel: C₁₂H₂₀N₅O₄P
MDL Number: MFCD07783825
CAS-nummer: 116384-53-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR