N,N-Diethylformamid ≥99.0% (by GC) for biochemical research

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Formic acid diethylamide, Formdiethylamide

D4234-100ML D4234-500ML
TCIAD4234-100MLEA 526 DKK
TCIAD4234-100ML TCIAD4234-500ML
N,N-Diethylformamid ≥99.0% (by GC) for biochemical research
N,N-Diethylformamid
Formel: C₅H₁₁NO
Kogepunkt: 177°C
Flammepunkt: 60°C
MDL Number: MFCD00003287
CAS-nummer: 617-84-5
EINECS: 210-533-2
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Suitability for use in ELISA tests to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR