5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D4183-1G D4183-5G
TCIAD4183-1GEA 2920 DKK
TCIAD4183-1G TCIAD4183-5G
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene ≥97.0% (by GC)
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene
Formel: C₂₀H₂₈Br₂S₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD23380207
CAS-nummer: 170702-05-3

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR