10-Deacetylbaccatin III ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D4148-500MG D4148-100MG
TCIAD4148-500MGEA 1690 DKK
TCIAD4148-500MG TCIAD4148-100MG
10-Deacetylbaccatin III ≥98.0% (by HPLC)
10-Deacetylbaccatin III
Formel: C₂₉H₃₆O₁₀
Molekylvægt: 544.60 g/mol
Smeltepunkt: 234°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00132913
CAS-nummer: 32981-86-5
EINECS: 418-680-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -40.0 to -44.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR