Doxycycline hyclate ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Doxycycline Hydrochloride Hemiethanolate Hemihydrate

D4116-5G D4116-25G
TCIAD4116-5GEA 214 DKK
TCIAD4116-5G TCIAD4116-25G
Doxycycline hyclate ≥97.0% (by HPLC)
Doxycycline hyclate
Formel: C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
Molekylvægt: 512.94 g/mol (503.94 anhy)
MDL Number: MFCD07357237
CAS-nummer: 24390-14-5
Merck Index: 14,03440

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D -100 to -107 deg (C=1, 0.01mol/L HCl (MeOH))
Water 1.4 to 2.8 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR