4,5-Difluoranthranilsyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2-Amino-4,5-difluorobenzoic acid
D4063-5G D4063-25G
TCIAD4063-5GEA 641 DKK
TCIAD4063-5G TCIAD4063-25G
4,5-Difluoranthranilsyre ≥98.0% (by HPLC, titration analysis)
4,5-Difluoranthranilsyre
Formel: C₇H₅F₂NO₂
Molekylvægt: 173.12 g/mol
Smeltepunkt: 180°C
MDL Number: MFCD00077509
CAS-nummer: 83506-93-8
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %