Diethylentriamin 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
114310010
114310025
114312500
114310100
ACRO114310010EA
647
DKK
ACRO114310010
ACRO114310025
ACRO114312500
ACRO114310100
Diethylentriamin 98+%
Diethylentriamin
Formel:
C₄H₁₃N₃ Kogepunkt: 206 °C (760 mmHg) Smeltepunkt: –40 °C Massefylde: 0,96 g/cm³ Flammepunkt: 96 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008171 CAS-nummer: 111-40-0 EINECS: 203-865-4 UN nummer: 2079 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4820 to 1.4850 (20°C, 589 nm) |
GC | ≥98.0 % |
Color scale | ≤30 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...