2,4-Dimethoxy-1,3,5-triazine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3998-1G
D3998-5G
TCIAD3998-1GEA
534
DKK
TCIAD3998-1G
TCIAD3998-5G
2,4-Dimethoxy-1,3,5-triazine ≥98.0% (by GC)
2,4-Dimethoxy-1,3,5-triazine
Formel:
C₅H₇N₃O₂ |
MDL Number:
MFCD20257908 CAS-nummer: 1898-72-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...