Dimethyl-2-(1,3-dithiole)phosphonate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(1,3-Dithiole)phosphonic Acid Dimethyl Ester

D3981-1G D3981-5G
TCIAD3981-1GEA 1680 DKK
TCIAD3981-1G TCIAD3981-5G
Dimethyl-2-(1,3-dithiole)phosphonate ≥97.0% (by GC)
Dimethyl-2-(1,3-dithiole)phosphonate
Formel: C₅H₉O₃PS₂
Opbevaringstemperatur: Fryser
CAS-nummer: 133113-76-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR