1,2-Dipalmitoyl-sn-glycero-3-phosphocholine ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,3-Bis(palmitoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, L-DPPC, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, L-α-Lecithin, Dipalmitoyl, L-alpha-Dipalmitoyl phosphatidylcholine

D3925-1G D3925-200MG
TCIAD3925-1GEA 2090 DKK
TCIAD3925-1G TCIAD3925-200MG
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine ≥98.0% (by HPLC)
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
Formel: C₄₀H₈₀NO₈P
Smeltepunkt: 220°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00036903
CAS-nummer: 63-89-8
EINECS: 200-567-6
Merck Index: 14,02477

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +5.0 to +8.0 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR