2,2'-Dipyrrolylmethane ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Di(1H-pyrrol-2-yl)methane

D3851-1G D3851-5G
TCIAD3851-1GEA 1220 DKK
TCIAD3851-1G TCIAD3851-5G
2,2'-Dipyrrolylmethane ≥97.0% (by GC)
2,2'-Dipyrrolylmethane
Formel: C₉H₁₀N₂
Kogepunkt: 120°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03931560
CAS-nummer: 21211-65-4

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 72.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR