N-(Diphenylmethylene)glycine ethyl ester ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate

D3833-25G D3833-5G
TCIAD3833-25GEA 452 DKK
TCIAD3833-25G TCIAD3833-5G
N-(Diphenylmethylene)glycine ethyl ester ≥97.0% (by titrimetric analysis)
N-(Diphenylmethylene)glycine ethyl ester
Formel: C₁₇H₁₇NO₂
Molekylvægt: 267.32 g/mol
Kogepunkt: 195°C
MDL Number: MFCD00010590
CAS-nummer: 69555-14-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR