1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D3812-1G D3812-5G D3812-25G D3812-100G
TCIAD3812-1GEA 271 DKK
TCIAD3812-1G TCIAD3812-5G TCIAD3812-25G TCIAD3812-100G
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate)
Formel: C₁₀H₈B₂F₁₀N₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01073546
CAS-nummer: 178439-26-4
UN nummer: 1759
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR