Diisopropyl allylboronate ≥95.0% (qNMR)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3738-1G
D3738-5G
TCIAD3738-1GEA
1060
DKK
TCIAD3738-1G
TCIAD3738-5G
Diisopropyl allylboronate ≥95.0% (qNMR)
Diisopropyl allylboronate
Formel:
C₉H₁₉BO₂ Opbevaringstemperatur: Køleskab |
CAS-nummer:
51851-79-7 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (1H-qNMR) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...