Dermatan sulphate sodium salt

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Chondroitin sulphate B sodium salt

D3672-100MG D3672-20MG
TCIAD3672-100MGEA 2410 DKK
TCIAD3672-100MG TCIAD3672-20MG
Dermatan sulphate sodium salt
Dermatan sulphate sodium salt
MDL Number: MFCD00151683
CAS-nummer: 54328-33-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Specific rotation [a]20/D -64.0 to -72.0 deg (C=1, H2O) (calcd.on dried substance)
Drying loss max. 10.0 %
Total Nitrogen 2.6 to 3.3 % (calcd.on dried substance)
Total Sulfur 5.7 to 6.5 % (calcd.on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR