3,3'-Dibromo-4,4'-dimethoxybiphenyl ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3603-1G D3603-5G
TCIAD3603-1GEA 673 DKK
TCIAD3603-1G TCIAD3603-5G
3,3'-Dibromo-4,4'-dimethoxybiphenyl ≥95.0% (by GC)
3,3'-Dibromo-4,4'-dimethoxybiphenyl
Formel: C₁₄H₁₂Br₂O₂
Molekylvægt: 372.06 g/mol
MDL Number: MFCD10000957
CAS-nummer: 108989-36-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR