2,5-Dibenzylidenecyclopentanone ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D3597-25G D3597-5G
TCIAD3597-25GEA 2630 DKK
TCIAD3597-25G TCIAD3597-5G
2,5-Dibenzylidenecyclopentanone ≥98.0% (by HPLC)
2,5-Dibenzylidenecyclopentanone
Formel: C₁₉H₁₆O
MDL Number: MFCD00019321
CAS-nummer: 895-80-7

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 190.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR