Dimethyl tetrafluorosuccinate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Perfluorosuccinic Acid Dimethyl Ester, Tetrafluorobutanedioic Acid Dimethyl Ester, Dimethyl Tetrafluorobutanedioate, Tetrafluorosuccinic Acid Dimethyl Ester, Dimethyl Perfluorosuccinate

D3588-1G D3588-5G
TCIAD3588-1GEA 165 DKK
TCIAD3588-1G TCIAD3588-5G
Dimethyl tetrafluorosuccinate ≥95.0% (by GC)
Dimethyl tetrafluorosuccinate
Formel: C₆H₆F₄O₄
Molekylvægt: 218.10 g/mol
Kogepunkt: 174°C
MDL Number: MFCD00043947
CAS-nummer: 356-36-5

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR