3,5-Dimethyl-1-adamantanemethanol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-(Hydroxymethyl)-3,5-dimethyladamantane
D3367-1G
D3367-5G
TCIAD3367-1GEA
501
DKK
TCIAD3367-1G
TCIAD3367-5G
3,5-Dimethyl-1-adamantanemethanol ≥98.0% (by GC)
3,5-Dimethyl-1-adamantanemethanol
Formel:
C₁₃H₂₂O Molekylvægt: 194.32 g/mol Kogepunkt: 115°C |
MDL Number:
MFCD00188062 CAS-nummer: 26919-42-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...