Dimesitylammonium pentafluorobenzenesulphonate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3293-1G D3293-5G
TCIAD3293-1GEA 755 DKK
TCIAD3293-1G TCIAD3293-5G
Dimesitylammonium pentafluorobenzenesulphonate ≥98.0% (by titrimetric analysis)
Dimesitylammonium pentafluorobenzenesulphonate
Formel: C₂₄H₂₄F₅NO₃S
Opbevaringstemperatur: Køleskab
CAS-nummer: 850629-65-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystaline
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR