Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3071-5G D3071-25G
TCIAD3071-5GEA 288 DKK
TCIAD3071-5G TCIAD3071-25G
Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)
Diethyl (bromodifluoromethyl)phosphonate
Formel: C₅H₁₀BrF₂O₃P
Molekylvægt: 267.01 g/mol
Kogepunkt: 102°C
MDL Number: MFCD00069118
CAS-nummer: 65094-22-6

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR