3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,6-Dibromo-9-phenyl-9H-carbazole

D2981-1G D2981-5G
TCIAD2981-1GEA 238 DKK
TCIAD2981-1G TCIAD2981-5G
3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
3,6-Dibromo-9-phenylcarbazole
Formel: C₁₈H₁₁Br₂N
Molekylvægt: 401.10 g/mol
MDL Number: MFCD08276338
CAS-nummer: 57103-20-5

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR