Diisopropyl sulphate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Diisopropyl sulphate

D2954-25G D2954-5G
TCIAD2954-25GEA 3070 DKK
TCIAD2954-25G TCIAD2954-5G
Diisopropyl sulphate ≥97.0% (by GC)
Diisopropyl sulphate
Formel: C₆H₁₄O₄S
Kogepunkt: 94°C
Smeltepunkt: -19°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD01682898
CAS-nummer: 2973-10-6
EINECS: 221-011-9
UN nummer: 1760
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR