1,1'-Dibutyrylferrocene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D2951-25G
D2951-5G
TCIAD2951-25GEA
2460
DKK
TCIAD2951-25G
TCIAD2951-5G
1,1'-Dibutyrylferrocene ≥98.0% (by GC)
1,1'-Dibutyrylferrocene
Formel:
C₁₈H₂₂FeO₂ Molekylvægt: 326.22 g/mol |
MDL Number:
MFCD01013566 CAS-nummer: 1274-06-2 |
Specifikationer Testresultater
Appearance | Amber to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...