(2,4-Dichlorobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Triphenyl(2,4-dichlorobenzyl)phosphonium Chloride

D2907-25G D2907-5G
TCIAD2907-25GEA 739 DKK
TCIAD2907-25G TCIAD2907-5G
(2,4-Dichlorobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(2,4-Dichlorobenzyl)triphenylphosphonium chloride
Formel: C₂₅H₂₀Cl₃P
Smeltepunkt: 249°C
MDL Number: MFCD00031641
CAS-nummer: 2492-23-1
EINECS: 219-659-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR