Diamantane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D2876-1G D2876-5G
TCIAD2876-1GEA 468 DKK
TCIAD2876-1G TCIAD2876-5G
Diamantane ≥98.0% (by GC)
Diamantane
Formel: C₁₄H₂₀
MDL Number: MFCD00171301
CAS-nummer: 2292-79-7
Merck Index: 14,02538

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 243.0 to 247.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR