8-Dimethylamino-1-octanol ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2846-1G D2846-5G
TCIAD2846-1GEA 559 DKK
TCIAD2846-1G TCIAD2846-5G
8-Dimethylamino-1-octanol ≥93.0% (by GC)
8-Dimethylamino-1-octanol
Formel: C₁₀H₂₃NO
Molekylvægt: 173.30 g/mol
MDL Number: MFCD03701100
CAS-nummer: 29823-87-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR