Ethyl-3-(dimethylamino)acrylate ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethyl 3-(dimethylamino)acrylate, 3-(Dimethylamino)acrylic Acid Ethyl Ester, Ethyl 3-(N,N-dimethylamino)acrylate

D2836-25G D2836-100G
TCIAD2836-25GEA 173 DKK
TCIAD2836-25G TCIAD2836-100G
Ethyl-3-(dimethylamino)acrylate ≥98.0% (by GC, titration analysis)
Ethyl-3-(dimethylamino)acrylate
Formel: C₇H₁₃NO₂
Molekylvægt: 143.19 g/mol
Kogepunkt: 136°C
Flammepunkt: 108°C
MDL Number: MFCD00144269
CAS-nummer: 924-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57f5e40f7d62e9047e28896eb442fd80c61378f5&vid=57f5e40f7d62e9047e28896eb442fd80c61378f5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR