1,5-Dichlor-1,1,3,3,5,5-hexamethyltrisiloxan ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D2825-5G
D2825-25G
TCIAD2825-5GEA
755
DKK
TCIAD2825-5G
TCIAD2825-25G
1,5-Dichlor-1,1,3,3,5,5-hexamethyltrisiloxan ≥98.0% (by GC, titration analysis)
1,5-Dichlor-1,1,3,3,5,5-hexamethyltrisiloxan
Formel:
C₆H₁₈Cl₂O₂Si₃ Kogepunkt: 184°C Flammepunkt: 76°C |
MDL Number:
MFCD00045147 CAS-nummer: 3582-71-6 EINECS: 222-707-5 UN nummer: 2987 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...