Dibenzo[b,e][1,4]dioxine ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2776-1G D2776-5G
TCIAD2776-1GEA 870 DKK
TCIAD2776-1G TCIAD2776-5G
Dibenzo[b,e][1,4]dioxine ≥99.0% (by GC)
Dibenzo[b,e][1,4]dioxine
Formel: C₁₂H₈O₂
MDL Number: MFCD00022282
CAS-nummer: 262-12-4
EINECS: 205-974-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 119.0 to 122.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR