3,3-Diphenyl-1-propanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D2760-25G D2760-5G
TCIAD2760-25GEA 5310 DKK
TCIAD2760-25G TCIAD2760-5G
3,3-Diphenyl-1-propanol ≥98.0% (by GC)
3,3-Diphenyl-1-propanol
Formel: C₁₅H₁₆O
Molekylvægt: 212.29 g/mol
Kogepunkt: 208°C
MDL Number: MFCD00002930
CAS-nummer: 20017-67-8
EINECS: 243-466-2

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR