Dimethylmalonyl dichloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylmalonyl Dichloride

D2723-25G D2723-5G
TCIAD2723-25GEA 2640 DKK
TCIAD2723-25G TCIAD2723-5G
Dimethylmalonyl dichloride ≥98.0% (by GC, titration analysis)
Dimethylmalonyl dichloride
Formel: C₅H₆Cl₂O₂
Molekylvægt: 169.00 g/mol
Flammepunkt: 51°C
MDL Number: MFCD00192081
CAS-nummer: 5659-93-8
UN nummer: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=742c275f5ccee4e7910379b0db2dbe2f6aca746a&vid=742c275f5ccee4e7910379b0db2dbe2f6aca746a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR