N,N'-Dimethylquinacridone ≥96.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
DMQA, 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione
D2687-1G
D2687-5G
TCIAD2687-1GEA
485
DKK
TCIAD2687-1G
TCIAD2687-5G
N,N'-Dimethylquinacridone ≥96.0% (by HPLC)
N,N'-Dimethylquinacridone
Formel:
C₂₂H₁₆N₂O₂ Molekylvægt: 340.38 g/mol |
MDL Number:
MFCD00683078 CAS-nummer: 19205-19-7 |
Specifikationer Testresultater
Appearance | Dark red to Brown powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Absorbance (E1%1cm) | min. 270 (H2SO4, 612 to 617nm) |
Absorbance (E1%1cm)2 | min. 230 (H2SO4, 564 to 569nm) |
Absorbance (E1%1cm)3 | min. 680 (H2SO4, 388 to 393nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...